Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTCCACTTTGGAAGAAACCTAT[C/G]GACATTGCGATCAGAATGGAGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC7 - coiled-coil domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001026383.2 | 616 | Missense Mutation | CGA,GGA | R,G 103 | NP_001021554.1 | |
NM_001321115.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | NP_001308044.1 | |
NM_145023.5 | 616 | Missense Mutation | CGA,GGA | R,G 103 | NP_659460.3 | |
XM_005252593.2 | 616 | Intron | XP_005252650.1 | |||
XM_006717505.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_006717568.1 | |
XM_006717506.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_006717569.1 | |
XM_011519663.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517965.1 | |
XM_011519664.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517966.1 | |
XM_011519666.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517968.1 | |
XM_011519667.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517969.1 | |
XM_011519668.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517970.1 | |
XM_011519669.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517971.1 | |
XM_011519670.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517972.1 | |
XM_011519671.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517973.1 | |
XM_011519672.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517974.1 | |
XM_011519673.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517975.1 | |
XM_011519674.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517976.1 | |
XM_011519675.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517977.1 | |
XM_011519676.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517978.1 | |
XM_011519677.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517979.1 | |
XM_011519678.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517980.1 | |
XM_011519679.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517981.1 | |
XM_011519680.2 | 616 | Intron | XP_011517982.1 | |||
XM_011519683.2 | 616 | Intron | XP_011517985.1 | |||
XM_011519684.2 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517986.1 | |
XM_011519685.1 | 616 | Intron | XP_011517987.1 | |||
XM_011519686.2 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517988.1 | |
XM_011519687.1 | 616 | Intron | XP_011517989.1 | |||
XM_011519688.2 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_011517990.1 | |
XM_017016638.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_016872127.1 | |
XM_017016639.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_016872128.1 | |
XM_017016640.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_016872129.1 | |
XM_017016641.1 | 616 | Intron | XP_016872130.1 | |||
XM_017016642.1 | 616 | Intron | XP_016872131.1 | |||
XM_017016643.1 | 616 | Intron | XP_016872132.1 | |||
XM_017016644.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_016872133.1 | |
XM_017016645.1 | 616 | Intron | XP_016872134.1 | |||
XM_017016646.1 | 616 | Intron | XP_016872135.1 | |||
XM_017016647.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_016872136.1 | |
XM_017016648.1 | 616 | Intron | XP_016872137.1 | |||
XM_017016649.1 | 616 | Intron | XP_016872138.1 | |||
XM_017016650.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_016872139.1 | |
XM_017016651.1 | 616 | Missense Mutation | CGA,GGA | R,G 103 | XP_016872140.1 | |
XM_017016652.1 | 616 | Intron | XP_016872141.1 | |||
XM_017016653.1 | 616 | Intron | XP_016872142.1 |
LOC101929431 - uncharacterized LOC101929431 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |