Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACCATCATACTATTGCCCTGTT[C/T]CTCATTTGCCTCATGAATAATAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600664 MIM: 611604 | ||||||||||||||||||||
Literature Links: |
CHUK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHUK - conserved helix-loop-helix ubiquitous kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278.4 | 2249 | Missense Mutation | AAA,GAA | K,E 730 | NP_001269.3 | |
NM_001320928.1 | 2249 | UTR 3 | NP_001307857.1 | |||
XM_017015611.1 | 2249 | Missense Mutation | AAA,GAA | K,E 730 | XP_016871100.1 | |
XM_017015612.1 | 2249 | Intron | XP_016871101.1 | |||
XM_017015613.1 | 2249 | Missense Mutation | AAA,GAA | K,E 326 | XP_016871102.1 |
ERLIN1 - ER lipid raft associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |