Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCTCAGCCTCTTTCTGGTGC[C/T]TGAGCTTCTGGTGCTTTTTCTATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RRP12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRP12 - ribosomal RNA processing 12 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145114.1 | 3726 | Missense Mutation | AAG,AGG | K,R 1135 | NP_001138586.1 | |
NM_001284337.1 | 3726 | Missense Mutation | AAG,AGG | K,R 1096 | NP_001271266.1 | |
NM_015179.3 | 3726 | Missense Mutation | AAG,AGG | K,R 1196 | NP_055994.2 | |
XM_011539555.1 | 3726 | Missense Mutation | AAG,AGG | K,R 1168 | XP_011537857.1 | |
XM_011539556.2 | 3726 | Intron | XP_011537858.1 |