Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGTGTTACCTGACAAATGAATA[A/G]CCCTTTTCTGGGAAAACTCTTATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603413 | ||||||||||||||||||||
Literature Links: |
TIAL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TIAL1 - TIA1 cytotoxic granule-associated RNA binding protein-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033925.1 | 1349 | Silent Mutation | GGC,GGT | G,G 258 | NP_001029097.1 | |
NM_001323964.1 | 1349 | Silent Mutation | GGC,GGT | G,G 118 | NP_001310893.1 | |
NM_001323965.1 | 1349 | Silent Mutation | GGC,GGT | G,G 118 | NP_001310894.1 | |
NM_001323967.1 | 1349 | Silent Mutation | GGC,GGT | G,G 118 | NP_001310896.1 | |
NM_001323968.1 | 1349 | Silent Mutation | GGC,GGT | G,G 219 | NP_001310897.1 | |
NM_001323969.1 | 1349 | Silent Mutation | GGC,GGT | G,G 202 | NP_001310898.1 | |
NM_001323970.1 | 1349 | Silent Mutation | GGC,GGT | G,G 219 | NP_001310899.1 | |
NM_003252.3 | 1349 | Silent Mutation | GGC,GGT | G,G 241 | NP_003243.1 | |
XM_017016605.1 | 1349 | Intron | XP_016872094.1 | |||
XM_017016606.1 | 1349 | Silent Mutation | GGC,GGT | G,G 118 | XP_016872095.1 |