Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTTTACATTTCCACTTAAGAG[C/T]GTTGTTTACTGTGGTAGGAACTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OBFC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OBFC1 - oligonucleotide/oligosaccharide binding fold containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024928.4 | Intron | NP_079204.2 | ||||
XM_006717976.3 | Intron | XP_006718039.1 | ||||
XM_011540184.2 | Intron | XP_011538486.1 | ||||
XM_017016669.1 | Intron | XP_016872158.1 | ||||
XM_017016670.1 | Intron | XP_016872159.1 |
SH3PXD2A - SH3 and PX domains 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |