Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACCCTTTTGGACATCATGAGGC[A/G]GTTCCGGGCTGAGATGGAGAAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HKDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HKDC1 - hexokinase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025130.3 | 258 | Missense Mutation | CAG,CGG | Q,R 42 | NP_079406.3 | |
XM_011540195.2 | 258 | Missense Mutation | CAG,CGG | Q,R 42 | XP_011538497.1 |
LOC101928994 - uncharacterized LOC101928994 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |