Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGTTGTTCCCGATAGAGTGAC[C/T]GCCTTCTGATCAGAGGTGGGAGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608407 MIM: 611198 | ||||||||||||||||||||
Literature Links: |
DPYSL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPYSL4 - dihydropyrimidinase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006426.2 | 51 | Missense Mutation | CGC,TGC | R,C 16 | NP_006417.2 | |
XM_005252658.3 | 51 | UTR 5 | XP_005252715.1 | |||
XM_011539125.2 | 51 | UTR 5 | XP_011537427.1 | |||
XM_017015482.1 | 51 | UTR 5 | XP_016870971.1 | |||
XM_017015483.1 | 51 | UTR 5 | XP_016870972.1 |
JAKMIP3 - Janus kinase and microtubule interacting protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |