Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTTTACTGAAATCTCAGAAGG[A/G]AGTTTTTTATTCACGCCATCGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604619 | ||||||||||||||||||||
Literature Links: |
LGI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LGI1 - leucine rich glioma inactivated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308275.1 | 591 | Silent Mutation | GGA,GGG | G,G 85 | NP_001295204.1 | |
NM_001308276.1 | 591 | Silent Mutation | GGA,GGG | G,G 85 | NP_001295205.1 | |
NM_005097.3 | 591 | Silent Mutation | GGA,GGG | G,G 85 | NP_005088.1 | |
XM_017016911.1 | 591 | Silent Mutation | GGA,GGG | G,G 85 | XP_016872400.1 | |
XM_017016912.1 | 591 | Silent Mutation | GGA,GGG | G,G 85 | XP_016872401.1 |
LOC101927032 - uncharacterized LOC101927032 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |