Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTGCACTCGCACTTTAGCTTAT[G/T]TGTAAAAAGGTGAGGTTCATATTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SKIDA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SKIDA1 - SKI/DACH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207371.3 | 3472 | Missense Mutation | AAT,CAT | N,H 705 | NP_997254.3 | |
XM_005252450.3 | 3472 | Missense Mutation | AAT,CAT | N,H 705 | XP_005252507.1 | |
XM_011519480.2 | 3472 | Missense Mutation | AAT,CAT | N,H 705 | XP_011517782.1 | |
XM_017016225.1 | 3472 | Missense Mutation | AAT,CAT | N,H 705 | XP_016871714.1 | |
XM_017016226.1 | 3472 | Missense Mutation | AAT,CAT | N,H 705 | XP_016871715.1 |