Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCAACCAGTCAGTTCCACCTGTG[C/T]CACCATTTGGGCAGCCCCAGCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607185 | ||||||||||||||||||||
Literature Links: |
SEC24C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC24C - SEC24 homolog C, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004922.3 | 420 | Missense Mutation | CCA,TCA | P,S 11 | NP_004913.2 | |
NM_198597.2 | 420 | Missense Mutation | CCA,TCA | P,S 11 | NP_940999.1 | |
XM_011540380.2 | 420 | Missense Mutation | CCA,TCA | P,S 11 | XP_011538682.1 | |
XM_011540381.2 | 420 | Missense Mutation | CCA,TCA | P,S 11 | XP_011538683.1 | |
XM_017016967.1 | 420 | UTR 5 | XP_016872456.1 | |||
XM_017016968.1 | 420 | UTR 5 | XP_016872457.1 |