Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGGGGCCGGTCCCGGAGGGCTC[A/G]ATCCGCATCTACAGCATGAGGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSTO1 - glutathione S-transferase omega 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191002.1 | 288 | Silent Mutation | TCA,TCG | S,S 23 | NP_001177931.1 | |
NM_001191003.1 | 288 | UTR 5 | NP_001177932.1 | |||
NM_004832.2 | 288 | Silent Mutation | TCA,TCG | S,S 23 | NP_004823.1 |