Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTTTCATACGCGCACGATCGA[A/G]AGCATCCTGGAGCCGGTGGCACAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 193065 | ||||||||||||||||||||
Literature Links: |
VCL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VCL - vinculin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003373.3 | 124 | Silent Mutation | GAA,GAG | E,E 10 | NP_003364.1 | |
NM_014000.2 | 124 | Silent Mutation | GAA,GAG | E,E 10 | NP_054706.1 |