Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGGACCGCCCGCCCCCTGGCC[G/T]GCAGAGGCCCGAGGACCGCTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC29A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC29A3 - solute carrier family 29 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174098.1 | 185 | Missense Mutation | CGG,CTG | R,L 43 | NP_001167569.1 | |
NM_018344.5 | 185 | Missense Mutation | CGG,CTG | R,L 43 | NP_060814.4 | |
XM_006717910.3 | 185 | UTR 5 | XP_006717973.1 | |||
XM_017016377.1 | 185 | UTR 5 | XP_016871866.1 | |||
XM_017016378.1 | 185 | UTR 5 | XP_016871867.1 |