Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTAGAAGCACTGAGAGATGCGGC[C/T]CCCTCGCAGGGTAAGGGGACCATCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSL5 - acyl-CoA synthetase long-chain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016234.3 | 241 | Silent Mutation | GCC,GCT | A,A 43 | NP_057318.2 | |
NM_203379.1 | 241 | Intron | NP_976313.1 | |||
NM_203380.1 | 241 | Intron | NP_976314.1 | |||
XM_017016324.1 | 241 | Silent Mutation | GCC,GCT | A,A 43 | XP_016871813.1 |