Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAAAAATAAAAAGTATAATTACC[C/T]GAATCACAGAATTCAAGTCTTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP43 - cilia and flagella associated protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025145.5 | 4166 | Missense Mutation | CAG,CGG | Q,R 1477 | NP_079421.5 | |
XM_005270171.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1478 | XP_005270228.1 | |
XM_005270172.3 | 4166 | Missense Mutation | CAG,CGG | Q,R 1449 | XP_005270229.1 | |
XM_011540196.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1516 | XP_011538498.1 | |
XM_011540197.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1487 | XP_011538499.1 | |
XM_011540198.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1477 | XP_011538500.1 | |
XM_011540199.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1477 | XP_011538501.1 | |
XM_011540200.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1350 | XP_011538502.1 | |
XM_011540201.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1516 | XP_011538503.1 | |
XM_011540202.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 1259 | XP_011538504.1 | |
XM_011540203.2 | 4166 | Missense Mutation | CAG,CGG | Q,R 777 | XP_011538505.1 | |
XM_017016681.1 | 4166 | Missense Mutation | CAG,CGG | Q,R 1515 | XP_016872170.1 | |
XM_017016682.1 | 4166 | Missense Mutation | CAG,CGG | Q,R 1400 | XP_016872171.1 | |
XM_017016683.1 | 4166 | Missense Mutation | CAG,CGG | Q,R 978 | XP_016872172.1 | |
XM_017016684.1 | 4166 | Intron | XP_016872173.1 |