Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCTCTTCCTCTTGTGTGCTCT[A/G]GGCTTGGGCAGGGACTTGCATTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611486 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYCE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYCE1 - synaptonemal complex central element protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143763.1 | 985 | Nonsense Mutation | CAG,TAG | Q,* 294 | NP_001137235.1 | |
NM_001143764.1 | 985 | Nonsense Mutation | CAG,TAG | Q,* 294 | NP_001137236.1 | |
NM_130784.2 | 985 | Nonsense Mutation | CAG,TAG | Q,* 258 | NP_570140.1 |