Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTGCCTTTAGCAAGTCTGACAT[C/T]ACAAGAGAATCATCTTTTACATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616464 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECD - ecdysoneless cell cycle regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM149B1 - family with sequence similarity 149 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173348.1 | 386 | Silent Mutation | ATC,ATT | I,I 55 | NP_775483.1 | |
XM_005269744.2 | 386 | Silent Mutation | ATC,ATT | I,I 55 | XP_005269801.1 | |
XM_005269745.2 | 386 | Silent Mutation | ATC,ATT | I,I 55 | XP_005269802.1 | |
XM_005269747.3 | 386 | UTR 5 | XP_005269804.1 | |||
XM_011539737.2 | 386 | Missense Mutation | TCA,TTA | S,L 34 | XP_011538039.1 | |
XM_011539740.2 | 386 | Silent Mutation | ATC,ATT | I,I 55 | XP_011538042.1 | |
XM_017016164.1 | 386 | Missense Mutation | TCA,TTA | S,L 34 | XP_016871653.1 | |
XM_017016165.1 | 386 | Intron | XP_016871654.1 | |||
XM_017016166.1 | 386 | Intron | XP_016871655.1 | |||
XM_017016167.1 | 386 | Silent Mutation | ATC,ATT | I,I 55 | XP_016871656.1 |