Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCTTGACAGCGTTGTCTGACC[A/G]GCACTGAGAGACAGACACCTGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609389 MIM: 610909 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5F - inositol polyphosphate-5-phosphatase F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCMBP - minichromosome maintenance complex binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256378.1 | 1961 | Silent Mutation | GCC,GCT | A,A 605 | NP_001243307.1 | |
NM_001256379.1 | 1961 | Silent Mutation | GCC,GCT | A,A 432 | NP_001243308.1 | |
NM_024834.3 | 1961 | Silent Mutation | GCC,GCT | A,A 607 | NP_079110.1 | |
XM_005270157.4 | 1961 | Silent Mutation | GCC,GCT | A,A 552 | XP_005270214.1 | |
XM_011540170.1 | 1961 | Silent Mutation | GCC,GCT | A,A 537 | XP_011538472.1 | |
XM_017016663.1 | 1961 | Silent Mutation | GCC,GCT | A,A 550 | XP_016872152.1 | |
XM_017016664.1 | 1961 | Silent Mutation | GCC,GCT | A,A 535 | XP_016872153.1 |