Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCTCCATGGAGAACCAGCCTAA[C/T]TGCACACAACACCCAAGGAGCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615389 MIM: 615391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTPBP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTPBP4 - GTP binding protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IDI2 - isopentenyl-diphosphate delta isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033261.2 | Intron | NP_150286.1 |
IDI2-AS1 - IDI2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |