Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCAGAACGTGCAGATTCTGCTC[C/G]AGGCCGCCAGCTACCTGGAGCAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MXI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MXI1 - MAX interactor 1, dimerization protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008541.1 | 436 | Intron | NP_001008541.1 | |||
NM_005962.4 | 436 | Intron | NP_005953.4 | |||
NM_130439.3 | 436 | Missense Mutation | CAG,GAG | Q,E 78 | NP_569157.2 |