Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAGAACTTCTTGTGTCAGGGTA[A/C]CTAGGTGAATACTGTTCGAGAGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176943 | ||||||||||||||||||||
Literature Links: |
FGFR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGFR2 - fibroblast growth factor receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000141.4 | 3280 | Missense Mutation | AGG,AGT | R,S 782 | NP_000132.3 | |
NM_001144913.1 | 3280 | Intron | NP_001138385.1 | |||
NM_001144914.1 | 3280 | Missense Mutation | AGG,AGT | R,S 670 | NP_001138386.1 | |
NM_001144915.1 | 3280 | Intron | NP_001138387.1 | |||
NM_001144916.1 | 3280 | Missense Mutation | AGG,AGT | R,S 667 | NP_001138388.1 | |
NM_001144917.1 | 3280 | Missense Mutation | AGG,AGT | R,S 666 | NP_001138389.1 | |
NM_001144918.1 | 3280 | Missense Mutation | AGG,AGT | R,S 665 | NP_001138390.1 | |
NM_001144919.1 | 3280 | Intron | NP_001138391.1 | |||
NM_001320654.1 | 3280 | Missense Mutation | AGG,AGT | R,S 554 | NP_001307583.1 | |
NM_001320658.1 | 3280 | Missense Mutation | AGG,AGT | R,S 780 | NP_001307587.1 | |
NM_022970.3 | 3280 | Missense Mutation | AGG,AGT | R,S 783 | NP_075259.4 | |
NM_023029.2 | 3280 | Missense Mutation | AGG,AGT | R,S 693 | NP_075418.1 | |
XM_006717708.2 | 3280 | Missense Mutation | AGG,AGT | R,S 800 | XP_006717771.1 | |
XM_006717710.3 | 3280 | UTR 3 | XP_006717773.1 | |||
XM_006717711.2 | 3280 | Missense Mutation | AGG,AGT | R,S 713 | XP_006717774.1 | |
XM_006717712.2 | 3280 | Missense Mutation | AGG,AGT | R,S 687 | XP_006717775.1 | |
XM_017015920.1 | 3280 | UTR 3 | XP_016871409.1 | |||
XM_017015921.1 | 3280 | UTR 3 | XP_016871410.1 | |||
XM_017015922.1 | 3280 | Missense Mutation | AGG,AGT | R,S 711 | XP_016871411.1 | |
XM_017015923.1 | 3280 | Missense Mutation | AGG,AGT | R,S 710 | XP_016871412.1 | |
XM_017015924.1 | 3280 | Missense Mutation | AGG,AGT | R,S 686 | XP_016871413.1 | |
XM_017015925.1 | 3280 | UTR 3 | XP_016871414.1 |