Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGACTTGGAGGCGCTCTGGGCCC[A/G]CTGAACAATGGCAGAACACTTCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613597 | ||||||||||||||||||||
Literature Links: |
HOGA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOGA1 - 4-hydroxy-2-oxoglutarate aldolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MORN4 - MORN repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098831.1 | 702 | Intron | NP_001092301.1 | |||
NM_178832.3 | 702 | Missense Mutation | CGG,TGG | R,W 133 | NP_849154.1 | |
XM_011539250.2 | 702 | Missense Mutation | CGG,TGG | R,W 191 | XP_011537552.1 | |
XM_011539251.2 | 702 | Missense Mutation | CGG,TGG | R,W 133 | XP_011537553.1 |