Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGTGCAGCTGGGGGCTTTCCTG[C/T]GATGAAGACAGCCGCGCAGGACCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609502 MIM: 616103 | ||||||||||||||||||||
Literature Links: |
CDHR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDHR1 - cadherin related family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRIT1 - leucine rich repeat, Ig-like and transmembrane domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRIT2 - leucine rich repeat, Ig-like and transmembrane domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017924.3 | 1577 | Missense Mutation | NP_001017924.1 | |||
NM_001284223.1 | 1577 | Missense Mutation | NP_001271152.1 |