Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGGAAATGGCTGCTTCATCATC[A/C]TCCTCCTCAGCTGGTGGGGTCAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613427 MIM: 614215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANAPC16 - anaphase promoting complex subunit 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242546.1 | 184 | Silent Mutation | TCA,TCC | S,S 6 | NP_001229475.1 | |
NM_001242547.1 | 184 | Silent Mutation | TCA,TCC | S,S 6 | NP_001229476.1 | |
NM_001242548.1 | 184 | Silent Mutation | TCA,TCC | S,S 6 | NP_001229477.1 | |
NM_173473.3 | 184 | Silent Mutation | TCA,TCC | S,S 6 | NP_775744.1 |
ASCC1 - activating signal cointegrator 1 complex subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |