Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAGGCACCATCAAAGTAGCCAA[A/G]GGTACAAGAGAAAGGCACGAGCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611910 | ||||||||||||||||||||
Literature Links: |
SLC16A12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC16A12 - solute carrier family 16 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213606.3 | 1336 | Missense Mutation | CTT,TTT | L,F 383 | NP_998771.3 | |
XM_017016237.1 | 1336 | Missense Mutation | CTT,TTT | L,F 383 | XP_016871726.1 | |
XM_017016238.1 | 1336 | Missense Mutation | CTT,TTT | L,F 383 | XP_016871727.1 | |
XM_017016239.1 | 1336 | Missense Mutation | CTT,TTT | L,F 383 | XP_016871728.1 |