Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTCCTGGCGAGCTCCTTCCGGG[C/T]CCGCTCCTCGGCCCCTGCGGGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602267 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM8 - ADAM metallopeptidase domain 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBGCP2 - tubulin gamma complex associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256617.1 | 2759 | Missense Mutation | ACC,GCC | T,A 824 | NP_001243546.1 | |
NM_001256618.1 | 2759 | Missense Mutation | ACC,GCC | T,A 666 | NP_001243547.1 | |
NM_006659.3 | 2759 | Missense Mutation | ACC,GCC | T,A 796 | NP_006650.1 | |
XM_017015595.1 | 2759 | Missense Mutation | ACC,GCC | T,A 405 | XP_016871084.1 | |
XM_017015596.1 | 2759 | Missense Mutation | ACC,GCC | T,A 405 | XP_016871085.1 |