Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCTTAACTCAGCTTTTCCTCC[C/T]AGTGCTAAGCATGCCTTCTCATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609783 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITIH5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITIH5 - inter-alpha-trypsin inhibitor heavy chain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001851.2 | 4285 | Intron | NP_001001851.1 | |||
NM_030569.6 | 4285 | UTR 3 | NP_085046.5 | |||
NM_032817.5 | 4285 | UTR 3 | NP_116206.4 | |||
XM_011519713.2 | 4285 | UTR 3 | XP_011518015.1 | |||
XM_011519714.2 | 4285 | Intron | XP_011518016.1 |