Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTGCATACTGTGCCCCAGCAGA[C/T]CCCCGCTTTGGCGTTGAAGGCGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606283 | ||||||||||||||||||||
Literature Links: |
SORCS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SORCS1 - sortilin related VPS10 domain containing receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013031.2 | 3823 | Intron | NP_001013049.1 | |||
NM_001206569.1 | 3823 | Intron | NP_001193498.1 | |||
NM_001206570.1 | 3823 | Intron | NP_001193499.1 | |||
NM_001206571.1 | 3823 | Intron | NP_001193500.1 | |||
NM_001206572.1 | 3823 | UTR 3 | NP_001193501.1 | |||
NM_052918.4 | 3823 | Silent Mutation | GGA,GGG | G,G 1160 | NP_443150.3 | |
XM_011539199.2 | 3823 | Intron | XP_011537501.1 | |||
XM_011539201.2 | 3823 | Intron | XP_011537503.1 | |||
XM_017015614.1 | 3823 | Intron | XP_016871103.1 | |||
XM_017015615.1 | 3823 | Intron | XP_016871104.1 | |||
XM_017015616.1 | 3823 | Intron | XP_016871105.1 | |||
XM_017015617.1 | 3823 | Intron | XP_016871106.1 | |||
XM_017015618.1 | 3823 | Intron | XP_016871107.1 |