Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGCGCCCTTCCTCTGGGTGGCT[A/G]CAGTTCAGCCTCTGGCCCTCCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604647 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALY - calcyon neuron specific vesicular protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321617.1 | 1425 | UTR 5 | NP_001308546.1 | |||
NM_015722.3 | 1425 | Silent Mutation | TGC,TGT | C,C 74 | NP_056537.1 | |
XM_005252685.4 | 1425 | Silent Mutation | TGC,TGT | C,C 74 | XP_005252742.2 | |
XM_011539836.2 | 1425 | Silent Mutation | TGC,TGT | C,C 74 | XP_011538138.1 | |
XM_011539838.2 | 1425 | UTR 5 | XP_011538140.1 |