Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGTGAGCCACTGTGCCCAACT[C/T]AGAGTTCATACTGTTAATGACTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605143 MIM: 607035 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTR1A - ARP1 actin-related protein 1 homolog A, centractin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUFU - SUFU negative regulator of hedgehog signaling | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178133.1 | Intron | NP_001171604.1 | ||||
NM_016169.3 | Intron | NP_057253.2 | ||||
XM_011539858.2 | Intron | XP_011538160.1 | ||||
XM_011539860.2 | Intron | XP_011538162.1 | ||||
XM_011539861.2 | Intron | XP_011538163.1 | ||||
XM_011539863.2 | Intron | XP_011538165.1 | ||||
XM_011539864.2 | Intron | XP_011538166.1 | ||||
XM_017016323.1 | Intron | XP_016871812.1 |