Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGCAGCGTCACCTGTAGTAGTCT[C/T]CTCCCAGCATGCCGATAGGCACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPS1 - HPS1, biogenesis of lysosomal organelles complex 3 subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000195.4 | 2119 | Missense Mutation | GAA,GGA | E,G 643 | NP_000186.2 | |
NM_001311345.1 | 2119 | Missense Mutation | GAA,GGA | E,G 319 | NP_001298274.1 | |
NM_001322476.1 | 2119 | Missense Mutation | GAA,GGA | E,G 643 | NP_001309405.1 | |
NM_001322477.1 | 2119 | Missense Mutation | GAA,GGA | E,G 643 | NP_001309406.1 | |
NM_001322478.1 | 2119 | Missense Mutation | GAA,GGA | E,G 610 | NP_001309407.1 | |
NM_001322479.1 | 2119 | Missense Mutation | GAA,GGA | E,G 610 | NP_001309408.1 | |
NM_001322480.1 | 2119 | Missense Mutation | GAA,GGA | E,G 556 | NP_001309409.1 | |
NM_001322481.1 | 2119 | Missense Mutation | GAA,GGA | E,G 556 | NP_001309410.1 | |
NM_001322482.1 | 2119 | Missense Mutation | GAA,GGA | E,G 523 | NP_001309411.1 | |
NM_001322483.1 | 2119 | Missense Mutation | GAA,GGA | E,G 520 | NP_001309412.1 | |
NM_001322484.1 | 2119 | Missense Mutation | GAA,GGA | E,G 520 | NP_001309413.1 | |
NM_001322485.1 | 2119 | Missense Mutation | GAA,GGA | E,G 487 | NP_001309414.1 | |
NM_001322487.1 | 2119 | Missense Mutation | GAA,GGA | E,G 319 | NP_001309416.1 | |
NM_001322489.1 | 2119 | Missense Mutation | GAA,GGA | E,G 319 | NP_001309418.1 | |
NM_001322490.1 | 2119 | Intron | NP_001309419.1 | |||
NM_001322491.1 | 2119 | Intron | NP_001309420.1 | |||
NM_001322492.1 | 2119 | Intron | NP_001309421.1 | |||
NM_182639.3 | 2119 | Intron | NP_872577.1 | |||
XM_005269757.4 | 2119 | Missense Mutation | GAA,GGA | E,G 643 | XP_005269814.1 | |
XM_017016170.1 | 2119 | Missense Mutation | GAA,GGA | E,G 523 | XP_016871659.1 | |
XM_017016171.1 | 2119 | Missense Mutation | GAA,GGA | E,G 487 | XP_016871660.1 | |
XM_017016172.1 | 2119 | Missense Mutation | GAA,GGA | E,G 319 | XP_016871661.1 | |
XM_017016173.1 | 2119 | Intron | XP_016871662.1 |
PYROXD2 - pyridine nucleotide-disulphide oxidoreductase domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |