Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACCTTCCACTTTGGAGACAAAG[A/C]CCAGACTCCGCTTAAGGTCAGAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609330 MIM: 611653 | ||||||||||||||||||||
Literature Links: |
OIT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OIT3 - oncoprotein induced transcript 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G12B - phospholipase A2 group XIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318124.1 | 580 | Missense Mutation | GGC,GTC | G,V 149 | NP_001305053.1 | |
NM_001318125.1 | 580 | Missense Mutation | GGC,GTC | G,V 59 | NP_001305054.1 | |
NM_032562.4 | 580 | Missense Mutation | GGC,GTC | G,V 149 | NP_115951.2 | |
XM_005270244.4 | 580 | Missense Mutation | GGC,GTC | G,V 145 | XP_005270301.1 |