Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACAGAGATGAAGCATGAACTTAC[A/G]TGCTGGATGAGCGTTTCTACAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP21 - Rho GTPase activating protein 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020824.3 | 4751 | Intron | NP_065875.3 | |||
XM_005252542.3 | 4751 | Intron | XP_005252599.1 | |||
XM_005252544.4 | 4751 | Silent Mutation | CAC,CAT | H,H 1324 | XP_005252601.1 | |
XM_011519602.2 | 4751 | Silent Mutation | CAC,CAT | H,H 1368 | XP_011517904.1 | |
XM_011519603.2 | 4751 | Silent Mutation | CAC,CAT | H,H 1368 | XP_011517905.1 | |
XM_011519604.2 | 4751 | Intron | XP_011517906.1 | |||
XM_011519605.2 | 4751 | Silent Mutation | CAC,CAT | H,H 1358 | XP_011517907.1 | |
XM_011519606.2 | 4751 | Silent Mutation | CAC,CAT | H,H 1300 | XP_011517908.1 | |
XM_011519607.2 | 4751 | Silent Mutation | CAC,CAT | H,H 1368 | XP_011517909.1 | |
XM_017016459.1 | 4751 | Silent Mutation | CAC,CAT | H,H 1334 | XP_016871948.1 | |
XM_017016460.1 | 4751 | Silent Mutation | CAC,CAT | H,H 1324 | XP_016871949.1 | |
XM_017016461.1 | 4751 | Intron | XP_016871950.1 |