Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACCGAATTTTTGCCACTTGTGC[A/G]ATCTTCTCTGCTTCTATAATCATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611604 | ||||||||||||||||||||
Literature Links: |
ERLIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERLIN1 - ER lipid raft associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100626.1 | 776 | Silent Mutation | ATC,ATT | I,I 223 | NP_001094096.1 | |
NM_006459.3 | 776 | Silent Mutation | ATC,ATT | I,I 223 | NP_006450.2 | |
XM_005269442.3 | 776 | Silent Mutation | ATC,ATT | I,I 223 | XP_005269499.1 | |
XM_017015541.1 | 776 | Silent Mutation | ATC,ATT | I,I 223 | XP_016871030.1 |