Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGTTCATGGATCACCGGGAGGT[C/G]CGAGTAGAGTGTTCTGAGGACACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611309 MIM: 610559 | ||||||||||||||||||||
Literature Links: |
C10orf10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C10orf10 - chromosome 10 open reading frame 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007021.3 | 903 | Missense Mutation | CAC,GAC | H,D 205 | NP_008952.1 |
RASSF4 - Ras association domain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032023.3 | 903 | Intron | NP_114412.2 | |||
XM_005271833.4 | 903 | Intron | XP_005271890.1 | |||
XM_005271835.4 | 903 | Intron | XP_005271892.1 | |||
XM_006718013.3 | 903 | Intron | XP_006718076.1 | |||
XM_006718014.3 | 903 | Intron | XP_006718077.1 | |||
XM_011540255.2 | 903 | Intron | XP_011538557.1 | |||
XM_017016754.1 | 903 | Intron | XP_016872243.1 | |||
XM_017016755.1 | 903 | Intron | XP_016872244.1 | |||
XM_017016756.1 | 903 | Intron | XP_016872245.1 | |||
XM_017016757.1 | 903 | Intron | XP_016872246.1 | |||
XM_017016758.1 | 903 | Intron | XP_016872247.1 |