Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGCGACGTCGGGGCCGAGCTAAC[A/C]CGGCCTCTGGACAAGAGTGAGTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605122 MIM: 605506 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUPV3L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUPV3L1 - Suv3 like RNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301683.1 | 343 | UTR 5 | NP_001288612.1 | |||
NM_001323584.1 | 343 | UTR 5 | NP_001310513.1 | |||
NM_001323585.1 | 343 | UTR 5 | NP_001310514.1 | |||
NM_001323586.1 | 343 | UTR 5 | NP_001310515.1 | |||
NM_001323587.1 | 343 | UTR 5 | NP_001310516.1 | |||
NM_001323588.1 | 343 | UTR 5 | NP_001310517.1 | |||
NM_003171.4 | 343 | Silent Mutation | ACA,ACC | T,T 85 | NP_003162.2 |
VPS26A - VPS26, retromer complex component A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |