Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGTAAAAAGATAAAGAAGGAA[A/G]TAGAGCCTCCTCCTAAGGATGTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611226 | ||||||||||||||||||||
Literature Links: |
ARMC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMC3 - armadillo repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282745.1 | 92 | Missense Mutation | ATA,GTA | I,V 9 | NP_001269674.1 | |
NM_001282746.1 | 92 | Missense Mutation | ATA,GTA | I,V 9 | NP_001269675.1 | |
NM_001282747.1 | 92 | UTR 5 | NP_001269676.1 | |||
NM_173081.4 | 92 | Missense Mutation | ATA,GTA | I,V 9 | NP_775104.2 | |
XM_005252380.2 | 92 | Missense Mutation | ATA,GTA | I,V 9 | XP_005252437.1 | |
XM_005252381.2 | 92 | UTR 5 | XP_005252438.1 | |||
XM_011519350.2 | 92 | Missense Mutation | ATA,GTA | I,V 9 | XP_011517652.1 | |
XM_011519351.2 | 92 | Intron | XP_011517653.1 | |||
XM_017015831.1 | 92 | Missense Mutation | ATA,GTA | I,V 9 | XP_016871320.1 | |
XM_017015832.1 | 92 | Missense Mutation | ATA,GTA | I,V 9 | XP_016871321.1 | |
XM_017015833.1 | 92 | Missense Mutation | ATA,GTA | I,V 9 | XP_016871322.1 |