Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACAGGCCTGGTGTGCTGAGTCC[A/G]CACATAGTAAGCATTCAAAGAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602432 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC3 - coiled-coil domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPTN - optineurin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008211.1 | 367 | UTR 5 | NP_001008212.1 | |||
NM_001008212.1 | 367 | UTR 5 | NP_001008213.1 | |||
NM_001008213.1 | 367 | UTR 5 | NP_001008214.1 | |||
NM_021980.4 | 367 | Intron | NP_068815.2 |