Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCGCCTTCCCCGTGTGCTACCC[C/G]CTGGGCCGCCTGCTGGACTGGGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607802 | ||||||||||||||||||||
Literature Links: |
CNNM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNNM1 - cyclin and CBS domain divalent metal cation transport mediator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020348.2 | 1414 | Silent Mutation | CCC,CCG | P,P 375 | NP_065081.2 | |
XM_005269692.3 | 1414 | Silent Mutation | CCC,CCG | P,P 375 | XP_005269749.2 | |
XM_011539629.1 | 1414 | Silent Mutation | CCC,CCG | P,P 375 | XP_011537931.1 | |
XM_011539630.1 | 1414 | Silent Mutation | CCC,CCG | P,P 375 | XP_011537932.1 | |
XM_011539631.2 | 1414 | Silent Mutation | CCC,CCG | P,P 375 | XP_011537933.1 |