Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGGGGGAGGCAGGTGGAGAAT[C/T]GGGCCAGTGAGCTCATGGCAAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610734 MIM: 616388 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD2 - ankyrin repeat domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129981.2 | 253 | UTR 5 | NP_001123453.1 | |||
NM_001291218.1 | 253 | Missense Mutation | CGG,TGG | R,W 82 | NP_001278147.1 | |
NM_001291219.1 | 253 | Missense Mutation | CGG,TGG | R,W 82 | NP_001278148.1 | |
NM_020349.3 | 253 | UTR 5 | NP_065082.2 |
UBTD1 - ubiquitin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |