Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGGCTATTGGTTTATAAAATCC[C/T]CTGACCTCTTGGGTTTTAAAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614242 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC16A9 - solute carrier family 16 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323977.1 | 3332 | UTR 3 | NP_001310906.1 | |||
NM_001323978.1 | 3332 | UTR 3 | NP_001310907.1 | |||
NM_001323979.1 | 3332 | UTR 3 | NP_001310908.1 | |||
NM_001323980.1 | 3332 | UTR 3 | NP_001310909.1 | |||
NM_001323981.1 | 3332 | UTR 3 | NP_001310910.1 | |||
NM_194298.2 | 3332 | UTR 3 | NP_919274.1 | |||
XM_017015883.1 | 3332 | UTR 3 | XP_016871372.1 | |||
XM_017015884.1 | 3332 | UTR 3 | XP_016871373.1 |