Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGAGCTCAATTTAGTAGAAAGAA[C/T]TGGTGTACCTGAAAGCATTTAACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606743 MIM: 616872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLL2 - tolloid like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM9SF3 - transmembrane 9 superfamily member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020123.3 | 4275 | UTR 3 | NP_064508.3 | |||
XM_011539976.2 | 4275 | Intron | XP_011538278.1 | |||
XM_011539977.1 | 4275 | UTR 3 | XP_011538279.1 |