Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTGGCAGGAGTTGGCATCATTG[C/G]TGTTCTTGATGGGGTGGCTGAGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600767 | ||||||||||||||||||||
Literature Links: |
FAM208B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM208B - family with sequence similarity 208 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GDI2 - GDP dissociation inhibitor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115156.1 | 918 | Missense Mutation | ACC,AGC | T,S 266 | NP_001108628.1 | |
NM_001494.3 | 918 | Missense Mutation | ACC,AGC | T,S 311 | NP_001485.2 | |
XM_017016071.1 | 918 | Missense Mutation | ACC,AGC | T,S 289 | XP_016871560.1 |