Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATTTCTTGGTCTTTACCCTAT[A/G]TAAGGCAAGGAGAAAAAGACATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601299 MIM: 608925 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMPR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMPR1A - bone morphogenetic protein receptor type 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMRN2 - multimerin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024756.2 | 4019 | UTR 3 | NP_079032.2 | |||
XM_005270153.1 | 4019 | UTR 3 | XP_005270210.1 | |||
XM_006717970.3 | 4019 | UTR 3 | XP_006718033.1 |