Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCAACGGAGGAGCTGAATGAA[A/G]AGAAAATCAAGCTGGAATCTAAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC172 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC172 - coiled-coil domain containing 172 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198515.2 | 721 | Missense Mutation | AAG,GAG | K,E 48 | NP_940917.1 | |
XM_006717820.3 | 721 | Missense Mutation | AAG,GAG | K,E 48 | XP_006717883.1 | |
XM_011539771.2 | 721 | Missense Mutation | AAG,GAG | K,E 48 | XP_011538073.1 | |
XM_017016206.1 | 721 | Missense Mutation | AAG,GAG | K,E 48 | XP_016871695.1 |