Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGTGTGGGGGATGTATCCACAC[A/G]GTTCCTGTTGTACATGTCTAGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606615 MIM: 602140 MIM: 610258 | ||||||||||||||||||||
Literature Links: |
HIF1AN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIF1AN - hypoxia inducible factor 1 alpha subunit inhibitor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFB8 - NADH:ubiquinone oxidoreductase subunit B8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284367.1 | 418 | Missense Mutation | CGT,TGT | R,C 116 | NP_001271296.1 | |
NM_001284368.1 | 418 | Intron | NP_001271297.1 | |||
NM_005004.3 | 418 | Missense Mutation | CGT,TGT | R,C 116 | NP_004995.1 |
SEC31B - SEC31 homolog B, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |