Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCCCTGAAAATTCTCCCCTGG[A/G]GCCCTGAGAAGATCGGCACCCGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604422 | ||||||||||||||||||||
Literature Links: |
PNLIPRP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PNLIPRP1 - pancreatic lipase related protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303135.1 | 169 | Missense Mutation | AGC,GGC | S,G 48 | NP_001290064.1 | |
NM_006229.3 | 169 | Missense Mutation | AGC,GGC | S,G 48 | NP_006220.1 | |
XM_011539868.1 | 169 | Missense Mutation | AGC,GGC | S,G 48 | XP_011538170.1 | |
XM_011539869.1 | 169 | UTR 5 | XP_011538171.1 |