Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGTTTTTTATTTATAGATATAT[C/G]AAGATGCTTATAGGGCGGAGTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600438 | ||||||||||||||||||||
Literature Links: |
TFAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TFAM - transcription factor A, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270782.1 | 703 | Missense Mutation | CAA,GAA | Q,E 100 | NP_001257711.1 | |
NM_003201.2 | 703 | Missense Mutation | CAA,GAA | Q,E 100 | NP_003192.1 | |
XM_011540120.2 | 703 | Missense Mutation | CAA,GAA | Q,E 100 | XP_011538422.1 | |
XM_011540121.2 | 703 | Missense Mutation | CAA,GAA | Q,E 100 | XP_011538423.1 |