Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTATGATGACGTCATGCCCGACGC[C/G]TTTCTCGCCGGGGCAAGAGCGCGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611310 | ||||||||||||||||||||
Literature Links: |
PSTK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSTK - phosphoseryl-tRNA kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153336.2 | 252 | Silent Mutation | GCC,GCG | A,A 62 | NP_699167.2 | |
XM_017015640.1 | 252 | Silent Mutation | GCC,GCG | A,A 62 | XP_016871129.1 | |
XM_017015641.1 | 252 | Silent Mutation | GCC,GCG | A,A 62 | XP_016871130.1 |