Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACAGAAGGTGGCTGTAGGCCTC[T/G]TAGTTCTGGGTCAGCCGCAGTTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607672 | ||||||||||||||||||||
Literature Links: |
CLCF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLCF1 - cardiotrophin-like cytokine factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166212.1 | 436 | Silent Mutation | NP_001159684.1 | |||
NM_013246.2 | 436 | Silent Mutation | NP_037378.1 |
LOC100130987 - uncharacterized LOC100130987 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |